Zinc acetate, anhydrous ≥99.9%

Supplier: Thermo Scientific Chemicals
Danger

H33734-09 H33734-18 H33734-30
AAH33734-09EA 62.27 USD
AAH33734-09 AAH33734-18 AAH33734-30
Zinc acetate, anhydrous ≥99.9%
Zinc acetate

Purity: 99.9+% CAS No.: 557-34-6 Molecular Formula: C4H6O4Zn Formula Weight: 183.47 Melting Point: 200° dec. UN No.: 3077 MDL No.: MFCD00012454

Formula: C₄H₆O₄Zn
MW: 183.48
Boiling Pt: 908
Melting Pt: 200 °C (dec.)
Density: 1.735
Storage Temperature: Ambient
MDL Number: MFCD00012454
CAS Number: 557-34-6
EINECS: 209-170-2
UN: 3077
ADR: 9 III
Merck Index: 12,10256

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR