Celite® 545, filter aid, treated with sodium carbonate, flux calcined

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DE, Diatomaceous earth, flux-calcined

H33152-0B H33152-0I
AAH33152-0BEA 85.78 USD
AAH33152-0B AAH33152-0I
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Celite® 545, filter aid

CAS No.: 68855-54-9 MDL No.: MFCD00132803

Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00132803
CAS Number: 68855-54-9
EINECS: 272-489-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR