Zinc chloride 0.7 M in THF
Supplier: Thermo Scientific Chemicals
|
Danger
|
H32962-AE
H32962-AP
AAH32962-AEEA
135.96
USD
AAH32962-AE
AAH32962-AP
Zinc chloride 0.7 M in THF
Zinc chloride
CAS No.: 7646-85-7
Molecular Formula: Cl2Zn
Formula Weight: 136.29
UN No.: 1993
MDL No.: MFCD00011295
Formula:
ZnCl₂ Density: 0.948 Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...