2-Amino-5-(trifluoromethoxy)benzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-amino-5-(trifluoromethoxy)benzoic acid

H32916-03 H32916-06
AAH32916-03EA 116 USD
AAH32916-03 AAH32916-06
2-Amino-5-(trifluoromethoxy)benzoic acid 98%
2-Amino-5-(trifluoromethoxy)benzoic acid

Purity: 98% CAS No.: 83265-56-9 Molecular Formula: C8H6F3NO3 Formula Weight: 221.14 MDL No.: MFCD00176651

Formula: C₈H₆F₃NO₃
MW: 221.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00176651
CAS Number: 83265-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR