(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27834-06
H27834-03
AAH27834-06EA
143
USD
AAH27834-06
AAH27834-03
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 99%
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol
Purity: 99%
CAS No.: 23364-44-5
Molecular Formula: C14H15NO
Formula Weight: 213.28
Melting Point: 141-143°
Optical Rotation: +6.9° (c=0.6 in ethanol)
Formula:
C₁₄H₁₅NO Melting Pt: 141…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074959 CAS Number: 23364-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...