Bis(tetramethylcyclopentadienyl)nickel(II) 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(tetramethylcyclopentadienyl)nickel

H27784-03 H27784-06
AAH27784-03EA 305 USD
AAH27784-03 AAH27784-06
Bis(tetramethylcyclopentadienyl)nickel(II) 98+%
Bis(tetramethylcyclopentadienyl)nickel (II)

Purity: 98+% CAS No.: 79019-60-6 Molecular Formula: C18H26Ni Formula Weight: 301.10 Storage Temperature: 2°C to 8°C Melting Point: 107-112° MDL No.: MFCD01862460

Formula: C₁₈H₂₆Ni
Melting Pt: 107…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01862460
CAS Number: 79019-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR