7-Bromo-2H-1,4-benzothiazin-3(4H)-one 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Bromo-2H-benzo[b][1,4]thiazin-3(4H)-one

H27563-03 H27563-06
AAH27563-03EA 108 USD
AAH27563-03 AAH27563-06
7-Bromo-2H-1,4-benzothiazin-3(4H)-one 97%
7-Bromo-2H-1,4-benzothiazin-3(4H)-one
Formula: C₈H₆BrNOS
MW: 244.11 g/mol
Melting Pt: 210…214 °C
Storage Temperature: Ambient
MDL Number: MFCD02660583
CAS Number: 90814-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR