Palladium 1% on carbon, granules, reduced

Supplier: Thermo Scientific Chemicals

046462-14 046462-36 046462-22
AA46462-22EA 411.99 USD
AA46462-22 AA46462-14 AA46462-36
Palladium 1% on carbon, granules, reduced
Palladium

3mm (0.13in) Pellets

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR