Lithium-tert-butanolate 99%

Supplier: Thermo Scientific Chemicals
Danger

046366-30 046366-A1
AA46366-30EA 238 USD
AA46366-30 AA46366-A1
Lithium-tert-butanolate 99%
Lithium tert-butanolate

Powder


Purity: 99% CAS No.: 1907-33-1 Molecular Formula: C4H9LiO Formula Weight: 80.06 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: White to cream Odor: Odorless Melting Point: ca 150° dec. Beilstein Reference: 3620018 UN No.: 3206 MDL No.: MFCD00050479

Formula: C₄H₉LiO
MW: 80.06 g/mol
Melting Pt: ∼ 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00050479
CAS Number: 1907-33-1
EINECS: 217-611-5
UN: 3206
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR