Potassium ≥99.95% (metals basis), solid
Supplier: Thermo Scientific Chemicals
|
Danger
|
Breakseal ampoule
045842-03
045842-06
AA45842-03EA
181.99
USD
AA45842-03
AA45842-06
Potassium ≥99.95% (metals basis), solid
Potassium
Formula:
K MW: 39.1 g/mol Boiling Pt: 760 °C (1013 hPa) Melting Pt: 63 °C Density: 0.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133776 CAS Number: 7440-09-7 EINECS: 231-119-8 UN: 2257 ADR: 4.3,I Merck Index: 12,07763 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...