Transition Metals, Plasma Standard Solution, Specpure®, Thermo Scientific

Supplier: Thermo Scientific Chemicals

044518-AE 044518-AP
AAA44518-AEEA 254.86 USD
AAA44518-AE AAA44518-AP
Transition Metals, Plasma Standard Solution, Specpure®, Thermo Scientific
Standards Cationic Standards Multi-Element Standards

Cd, Co, Cr, Cu, Fe, Mn, Ni, V, Zn @ 100 µg/ml


Molecular Formula: Matrix: 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00151266

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR