JM-21 Multi-element Oil Based Standards, Specpure®, Thermo Scientific
Supplier: Thermo Scientific Chemicals
036762-22
036769-22
AAA36762-22EA
198.85
USD
AAA36762-22
AAA36769-22
JM-21 Multi-element Oil Based Standards, Specpure®, Thermo Scientific
Standards
Cationic Standards
Multi-Element Standards
21 elements: Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, N, Ni, P, Pb, Sn, Ti, V, Zn
- Available at 100 µg/g (±5%) or 900 µg/g (±2%)
Molecular Formula: Matrix: Hydrocarbon oil
Physical Form: Liquid
MDL No.: MFCD00151262
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...