TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure, Thermo Scientific

Supplier: Thermo Fisher Scientific

J75892-K2 J75892-K8 J75892-AE
101170-656EA 164.49 USD
101170-656 101174-872 101171-426
TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

Used in Western blotting protocols.


  • Contaminant Testing:
  • DNase (endo): Passes test
  • DNase (exo): Passes test
  • RNase: Passes test
  • Protease: Passes test
  • pH(25°C, 20X): 7.4 ± 0.1


20X TBS (Tris Buffered Saline) solution consisting of 500mM Tris, 60mM KCl and 2.8M NaCl in high purity dH2O, pH 7.4.


Solution is 0.2μm filtered and dispensed into durable, square Nalgene bottles. The 5L size is supplied in an upright carton with a convenient dispensing spout.


Dilute to 1X with high purity water for a solution consisting of 25mM Tris, pH 7.4, 3.0mM KCl and 140mM NaCl.


Storage temperature of 20X solution: Ambient

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR