Water, Ultrapure for molecular biology nuclease-free
Supplier: Thermo Fisher Scientific
Synonyms:
Dihydrogen oxide
J71786-K2
J71786-AP
J71786-AE
J71786-K8
101175-318EA
76.15
USD
101175-318
101170-298
101174-104
101175-828
Water, Ultrapure for molecular biology nuclease-free
Water
High purity dH2O tested for contaminating nuclease activity. Product is 0.2 μm filtered and dispensed into vials and/or durable, square Nalgene bottles. The 5L size is supplied in an upright carton with a convenient dispensing spout.
Contaminant Testing:
RNase: Passes test
DNase (endo): Passes test
DNase (exo): Passes test
Protease: Passes test
Formula:
H₂O Boiling Pt: 100 °C Melting Pt: 0 °C Density: 1.000 Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 14,10039 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...