Vinylferrocene ≥97.0%

Supplier: TCI America

V0064-1G
TCV0064-1GEA 101.36 USD
TCV0064-1G
Vinylferrocene ≥97.0%
Vinylferrocene

CAS Number: 1271-51-8
MDL Number: MFCD00001434
Molecular Formula: C12H12Fe
Molecular Weight: 212.07
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Color: Yellow
Flash Point (°C): 62

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR