1H,1H,2H,2H-Perfluorooctyltrichlorosilane ≥97.0%

Supplier: TCI America

Synonyms: Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-trichlorosilane

T2577-5G
TCT2577-5GEA 115.58 USD
TCT2577-5G
1H,1H,2H,2H-Perfluorooctyltrichlorosilane ≥97.0%
1H,1H,2H,2H-Perfluorooctyltrichlorosilane

CAS Number: 78560-45-9
MDL Number: MFCD00042363
Molecular Formula: C8H4Cl3F13Si
Molecular Weight: 481.53
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 85
Flash Point (°C): 54
Specific Gravity (20/20): 1.65


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Trichloro(1H,1H,2H,2H-perfluoro-n-octyl)silane, 1-(Trichlorosilyl)-1H,1H,2H,2H-tridecafluoro-n-octane

Formula: C₈H₄Cl₃F₁₃Si
Boiling Pt: 85 °C
Density: 1.65 (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00042363
CAS Number: 78560-45-9
UN: 2987
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/62/91/28996291.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR