3a,4,7,7a-Tetrahydroindene ≥98.0% stabilised
Supplier: TCI America
T1315-500ML
T1315-25ML
TCT1315-500MLEA
530
USD
TCT1315-500ML
TCT1315-025ML
3a,4,7,7a-Tetrahydroindene ≥98.0% stabilised
3a,4,7,7a-Tetrahydroindene
(stabilized with BHT)
CAS Number: 3048-65-5
MDL Number: MFCD00021227
Molecular Formula: C9H12
Molecular Weight: 120.20
Purity/Analysis Method:
Formula:
C₉H₁₂ Boiling Pt: 160 °C Density: 0.93 (20 °C) Flash Pt: 38 °C |
MDL Number:
MFCD00021227 CAS Number: 3048-65-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...