5-Sulfosalicylic acid dihydrate ≥98.0% (by HPLC, titration analysis) for biochemical research
Supplier: TCI America
[for Biochemical Research]
CAS Number: 5965-83-3
MDL Number: MFCD00149540
Molecular Formula: C7H6O6S
Molecular Weight: 218.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 113
Lambda max.: 304 nm (EtOH)
Product Categories: Bioscience, Materials for Clinical Chemistry (Micellaneous), Deproteinization Agents
Other Product Names / Synonyms: 2-Hydroxy-5-sulfobenzoic Acid
Formula:
HO₃SC₆H₃-2-(OH)CO₂H·2H₂O Melting Pt: 113 °C |
MDL Number:
MFCD00149540 CAS Number: 5965-83-3 UN: 2585 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...