Glycerine ≥99.0%

Supplier: TCI America

Synonyms: Glyceritol, Glysanin, 1,2,3-Propanetriol, Osmoglyn, Glycerin, Glycyl alcohol, Glyrol, Propane-1,2,3-triol, Propanetriol, Trihydroxypropane, Glycerol

S0373-10G S0373-1G
TCS0373-10GEA 111 USD
TCS0373-10G TCS0373-1G
Glycerine ≥99.0%
Glycerine

[Matrix for FABMS and liquid SIMS] CAS Number: 56-81-5 MDL Number: MFCD00004722 Molecular Formula: C3H8O3 Molecular Weight: 92.09 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 290 Flash Point (°C): 177 Specific Gravity (20/20): 1.26


Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials Other Product Names / Synonyms: Glycerin

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C
Melting Pt: 18.6
Density: 1.26 (20 °C)
Flash Pt: 177 °C
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebb76b9cbe15a9472e70f24bba0cf4db0f10dc14&vid=1581f4bc25baf783b19fdaff0b00b76e5ab15b28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR