(4-Nitrophenyl)-α-D-glucopyranoside

Supplier: TCI America

Synonyms: 4-Nitrophenyl α-D-glucopyranoside(2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol4-Nitrophenyl a-D-glucopyranoside

N0493-5G N0493-1G
TCN0493-005GEA 164.57 USD
TCN0493-005G TCN0493-001G
(4-Nitrophenyl)-α-D-glucopyranoside
(4-Nitrophenyl) α-D-glucopyranoside

[Substrate for alpha-D-Galactosidase]
CAS Number: 3767-28-0
MDL Number: MFCD00064088
Molecular Formula: C12H15NO8
Molecular Weight: 301.25
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 212
Specific rotation [a]20/D: 218.5 deg (C=0.5, H2O)
Storage Temperature: 0-10°C

Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Melting Pt: 212 °C
MDL Number: MFCD00064088
CAS Number: 3767-28-0
https://us.vwr.com/stibo/web/std.lang.all/96/51/28989651.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR