1H,1H,9H-Hexadecafluoro-1-nonanol ≥97.0%
Supplier: TCI America
Synonyms:
1H,1H,9H-Perfluoro-1-nonanol
, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluor-1-nonanol
, 1H,1h,9h-hexadecafluorononanol
, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1-nonanol
CAS Number: 376-18-1
MDL Number: MFCD00039629
Molecular Formula: C9H4F16O
Molecular Weight: 432.10
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 59
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 1H,1H,9H-Perfluoro-1-nonanol
Formula:
C₉H₄F₁₆O MW: 432.1 g/mol Melting Pt: 59 °C |
MDL Number:
MFCD00039629 CAS Number: 376-18-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...