Hydrobromic acid 10 - 20% in ethanol for esterification

Supplier: TCI America
Danger

Synonyms: Hydrogen bromide

H0959-25ML H0959-500ML H0959-100ML
TCH0959-500MLEA 283.19 USD
TCH0959-500ML TCH0959-100ML TCH0959-25ML
Hydrobromic acid 10 - 20% in ethanol for esterification
Hydrogen bromide

[for Esterification]
CAS Number: 10035-10-6
MDL Number: MFCD00011323
Molecular Formula: HBr
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 16
Storage Temperature: 0-10°C

Formula: HBr
MW: 80.91 g/mol
Flash Pt: 16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Concentration (Neutralization titration) 10.0 to 20.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR