15-Hydroxypentadecanoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
H0676-5G
H0676-1G
TCH0676-5GEA
169.51
USD
TCH0676-5G
TCH0676-001G
15-Hydroxypentadecanoic acid ≥98.0% (by GC, titration analysis)
15-Hydroxypentadecanoic acid
CAS Number: 4617-33-8
MDL Number: MFCD00039535
Molecular Formula: C15H30O3
Molecular Weight: 258.40
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 88
Formula:
C₁₅H₃₀O₃ MW: 258.40 g/mol Melting Pt: 88 °C |
MDL Number:
MFCD00039535 CAS Number: 4617-33-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...