Hydrobromic acid 30% (w/w) ∼5.1 mol/L in acetic acid for peptide research

Supplier: TCI America
Danger

Synonyms: Anhydrous hydrobromic acidHydrogen bromide

H0182-25G H0182-500G H0182-100G
TCH0182-100GEA 32 USD
TCH0182-100G TCH0182-500G TCH0182-25G
Hydrobromic acid 30% (w/w) ∼5.1 mol/L in acetic acid for peptide research
Hydrogen bromide

[for Peptide research]
CAS Number: 10035-10-6
MDL Number: MFCD00011323
Molecular Formula: HBr
Molecular Weight: 80.91
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.40


Other Product Names / Synonyms: Hydrobromic Acid

Formula: HBr
MW: 80.91 g/mol
Density: 1.40 (20 °C)
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 2920
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/02/30/28970230.jpg

Order Now

Specification Test Results

Appearance Colourless to Red to Green clear liquid
Concentration (Argentmetric titration) 30.0 to 33.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR