Tartrazine
Supplier: TCI America
Synonyms:
Filter Yellow
F0144-25G
TCF0144-025GEA
119.57
USD
TCF0144-025G
Tartrazine
Tartrazine
CAS Number: 1934-21-0
MDL Number: MFCD00148908
Molecular Formula: C16H12N4O9S2
Molecular Weight: 534.36
Form: Powder
Color: Pale Yellow
Formula:
C₁₆H₁₂N₄O₉S₂ MW: 534.36 g/mol |
MDL Number:
MFCD00148908 CAS Number: 1934-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...