(R)-(+)-α,α-Diphenylprolinol ≥98.0%

Supplier: TCI America

Synonyms: (R)-(+)-alpha,alpha-Diphenylprolinol, (R)-(+)-α,α-Diphenyl-2-pyrrolidinemethanol, (R)-(+)-2-(Diphenylhydroxymethyl)pyrrolidine, α,α-Diphenyl-D-prolinol

D2365-5G D2365-1G
TCD2365-5GEA 426.8 USD
TCD2365-5G TCD2365-001G
(R)-(+)-α,α-Diphenylprolinol ≥98.0%
(R)-(+)-α,α-Diphenylprolinol

CAS Number: 22348-32-9 MDL Number: MFCD00077754 Molecular Formula: C17H19NO Molecular Weight: 253.35 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 78 Specific rotation [a]20/D: 58 deg (C=2, MeOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts Other Product Names / Synonyms: alpha,alpha-Diphenyl-D-prolinol, (R)-(+)-2-(Diphenylhydroxymethyl)pyrrolidine

Formula: C₁₇H₁₉NO
MW: 253.35 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00077754
CAS Number: 22348-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8970c5ef82d503856df88a822a352946d3bd8fa&vid=b8970c5ef82d503856df88a822a352946d3bd8fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR