cis-2,6-Dimethylmorpholine ≥97.0%
Supplier: TCI America
D0746-25ML
D0746-500ML
TCD0746-025MLEA
75
USD
TCD0746-025ML
TCD0746-500ML
cis-2,6-Dimethylmorpholine ≥97.0%
cis-2,6-Dimethylmorpholine
CAS Number: 6485-55-8
MDL Number: MFCD00078428
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method:
Formula:
C₆H₁₃NO Boiling Pt: 147 °C Density: 0.93 (20 °C) Flash Pt: 43 °C |
MDL Number:
MFCD00078428 CAS Number: 6485-55-8 UN: 1992 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...