3-Cyclohexene-1-carboxylic acid ≥98.0%
Supplier: TCI America
Synonyms:
Cyclohex-3-enecarboxylic acid, 3-Cyclohexenecarboxylic acid, 1,2,3,6-Tetrahydrobenzoic Acid
C0652-500G
C0652-25G
TCC0652-500GEA
265.7
USD
TCC0652-500G
TCC0652-25G
3-Cyclohexene-1-carboxylic acid ≥98.0%
3-Cyclohexene-1-carboxylic acid
CAS Number: 4771-80-6
MDL Number: MFCD00013781
Molecular Formula: C7H10O2
Molecular Weight: 126.16
Purity/Analysis Method:
Other Product Names / Synonyms: 1,2,3,6-Tetrahydrobenzoic Acid
Formula:
C₇H₁₀O₂ MW: 126.16 g/mol Boiling Pt: 238 °C Density: 1.08 (20 °C) |
MDL Number:
MFCD00013781 CAS Number: 4771-80-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...