N(α)-Cbz-L-tryptophan ≥98.0%

Supplier: TCI America

Synonyms: (S)-2-(((Benzyloxy)carbonyl)amino)-3-(1H-indol-3-yl)propanoic acid, N(α)-Benzyloxycarbonyl-L-tryptophan, N-Cbz-L-Tryptophan, Z-Trp-OH

C0638-5G C0638-25G
TCC0638-005GEA 23 USD
TCC0638-005G TCC0638-025G
N(α)-Cbz-L-tryptophan ≥98.0%
N(α)-Cbz-L-tryptophan

CAS Number: 7432-21-5 MDL Number: MFCD00065700 Molecular Formula: C19H18N2O4 Molecular Weight: 338.36 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 125 Specific rotation [a]20/D: 3 deg (C=3, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected) Other Product Names / Synonyms: N-Cbz-L-tryptophan, Z-Trp-OH

Formula: C₁₉H₁₈N₂O₄
MW: 338.4 g/mol
Melting Pt: 125 °C
MDL Number: MFCD00065700
CAS Number: 7432-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15336c3c00dae1f6e93f62bf8e45c823de055ec2&vid=15336c3c00dae1f6e93f62bf8e45c823de055ec2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR