(+/-)-ɑ-Bisabolol ≥90.0%

Supplier: TCI America

Synonyms: 6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol

B2119-25ML
TCB2119-025MLEA 83 USD
TCB2119-025ML
(+/-)-ɑ-Bisabolol ≥90.0%
(+/-)-alpha-Bisabolol

CAS Number: 515-69-5
MDL Number: MFCD00129082
Molecular Formula: C15H26O
Molecular Weight: 222.37
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Flash Point (°C): 135
Specific Gravity (20/20): 0.93


Product Categories: Bioscience, Sesquiterpenes
Other Product Names / Synonyms: 6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol

Formula: C₁₅H₂₆O
MW: 222.37 g/mol
Density: 0.93 (20 °C)
Flash Pt: 135 °C
MDL Number: MFCD00129082
CAS Number: 515-69-5
https://us.vwr.com/stibo/web/std.lang.all/38/93/28933893.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR