3,3'-Oxydipropionitrile ≥98.0%
Supplier: TCI America
Synonyms:
Bis(2-cyanoethyl) Ether
B0475-500ML
B0475-25ML
TCB0475-500MLEA
1111
USD
TCB0475-500ML
TCB0475-025ML
3,3'-Oxydipropionitrile ≥98.0%
3,3'-Oxydipropionitrile
CAS Number: 1656-48-0
MDL Number: MFCD00001960
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method:
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Bis(2-cyanoethyl) Ether
Formula:
C₆H₈N₂O Boiling Pt: 172 °C Density: 1.05 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00001960 CAS Number: 1656-48-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...