Benzyl methyl ether ≥97.0%

Supplier: TCI America

Synonyms: (Methoxymethyl)benzene

B0424-25ML B0424-500ML
TCB0424-025MLEA 69 USD
TCB0424-025ML TCB0424-500ML
Benzyl methyl ether ≥97.0%
Benzyl methyl ether

CAS Number: 538-86-3 MDL Number: MFCD00025901 Molecular Formula: C8H10O Molecular Weight: 122.17 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 174 Flash Point (°C): 56 Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Methoxymethylbenzene

Formula: C₈H₁₀O
Boiling Pt: 174 °C
Density: 0.97 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00025901
CAS Number: 538-86-3
UN: 3271
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR