5-Aminosalicylic acid ≥98.0% (by titrimetric analysis) for biochemical research
Supplier: TCI America
A2291-25G
A2291-5G
TCA2291-25GEA
115.71
USD
TCA2291-25G
TCA2291-5G
5-Aminosalicylic acid ≥98.0% (by titrimetric analysis) for biochemical research
5-Aminosalicylic acid
[for Biochemical Research]
CAS Number: 89-57-6
MDL Number: MFCD00007877
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 265
Storage Temperature: 0-10°C
Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 5-Amino-2-hydroxybenzoic Acid, 5-ASA
Formula:
C₇H₇NO₃ Melting Pt: 265 °C |
MDL Number:
MFCD00007877 CAS Number: 89-57-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...