6-Aminocaproic acid ≥98.0% (by titrimetric analysis) for Biochemical Research

Supplier: TCI America

Synonyms: ε-Amino-n-caproic acidH-Eahx-OHAminocaproic Acid6-Aminocaproic Acid6-Aminohexanoic acid

A2255-5G A2255-25G
TCA2255-5GEA 25.38 USD
TCA2255-5G TCA2255-25G
6-Aminocaproic acid ≥98.0% (by titrimetric analysis) for Biochemical Research
6-Aminocaproic acid

[for Biochemical Research]
CAS Number: 60-32-2
MDL Number: MFCD00008238
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 204
Storage Temperature: 0-10°C

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 204 °C
MDL Number: MFCD00008238
CAS Number: 60-32-2
https://us.vwr.com/stibo/web/std.lang.all/04/30/28930430.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR