Low-Pressure Chromatography Tubing, Bio-Rad
Supplier: Bio-Rad
7318212 7318207 7318247 7318208 7318213 7318214 7318242 7318241 7318209 7318210 7318211 7318215 7318240 7318249 7318248
101646-368EA 246.15 USD
101646-368 101646-464 101646-358 101646-406 101646-360 101646-370 101646-372 101646-404 101646-402 101646-362 101646-364 101646-366 101646-374 101646-400 101646-410 101646-408
Low-Pressure Chromatography Tubing, Bio-Rad
Tubing Chromatography Tubing
Bio-Rad supplies low-pressure tubing for most liquid-handling needs - both individually and as pump tubing kits.
  • Silicone tubing is recommended when biocompatibility is important
  • Material contains no cytotoxic extractables and has excellent wetting properties
  • Autoclavable and may be heated for pyrogen removal. It may be damaged by concentrated acids and bases
  • Tygon tubing is clear and tough, and will tolerate greater pressures than silicone
  • Transparency makes it ideal for plumbing chromatography systems
However, it should not be autoclaved, and may be damaged by high concentrations of alcohol. PharMed tubing has a wider range of chemical compatibility than silicone or Tygon and is the ideal tubing material for use in the pumpheads of the Model EP-1 Econo pump, Econo gradient pump, and BioLogic LP system.
PharMed tubing generally lasts 10 times longer than Tygon or silicone. PTFE tubing is chemically inert; it can be used with virtually any reagent, and is stable to 400 °C.