Protexis™ Latex Classic Sterile Powder-Free Gloves, Cardinal
Supplier: Cardinal
2D72N85X 2D72N65X 2D72N90X 2D72N75X 2D72N70X 2D72N55X 2D72N60X 2D72N80X
89237-230CS 2271.06 USD
89237-230 89237-228 89237-226 89237-224 89237-222 89237-234 89237-220 89237-232
Protexis™ Latex Classic Sterile Powder-Free Gloves, Cardinal
Gloves
Sterile Latex surgical gloves with nitrile coating delivers protection, dexterity and an advanced grip.
  • Exclusive glove design
  • Natural movement in the fingers and thumb
  • Interlocking, beaded cuff resists roll-down
Sterilized by gamma radiation to a sterility assurance level (SAL) of 10⁻⁶.
Ordering information: Natural color. 50 pairs/box; 4 boxes/case.