Fitting, LDPE, Straight, Hosebarb x Quick-Disconnect Couplers, 3/16" ID x 5/16" ID; 100/Pk
Supplier: VWR International
VWR Item#MFLX06290-09BLK
Your Organization's Price:
Retrieving
Provides easy single-handed disconnection.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/16" ID x 5/16" ID
Construction from low-density polyethylene (LDPE).