Smiths Medical® Fitting, Polycarbonate, One-Way Stopcock with Male Slip Luer, Non-Sterile; 1000/Pk
Supplier: ICU MEDICAL, INC.
Regulate flow without any leaking.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- One-way stopcock/flow diverter with male luer slip connection
- HDPE handle rotates 90°
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.