ChemFluor PTFE Tubing, 5/8" ID×3/4" OD; 50 ft/Pk
Supplier: VWR International
Be more flexible.
- Ideal for general-purpose, single-use, and extended fluid path applications
- 5/8 x 3/4" tubing size
<B>Applications:</B> Chemical transfer and pharmaceutical processes.<br /><B>Characteristics:</B> Most flexible fluoropolymer. Excellent chemical compatibility. Translucent.<br /><B>Certification:</B> FDA compliant.<br /><B>Temperature range:</B> –450 to 550 °F (–268 to 288 °C).<br /><B>Sterilize:</B> Autoclave, ethylene oxide, or dry heat.