McMillan Flow High-Accuracy Flowmeter, Stainless Steel, 0 to 5 VDC Output, 20 to 200 mL/min
Supplier: VWR International
VWR Item#MFLX32718-02
Your Organization's Price:
Retrieving
Economical design with an outstanding ±1.0 % full-scale accuracy.
  • A wide range of outputs options for integration into any system
Select these flowmeters and transmitters for extremely low flow-rate applications or for higher flow rate measurements and yet maintain a full-scale accuracy of ±1% or better. All units feature a stainless steel housing and come standard with stainless steel compression fittings.<br /><br />The flowmeters/transmitters can be used with a wide variety of transparent, low-viscosity (under 10 cSt) corrosive or neutral liquids and solvents. <br /><br /><B>Note:</B> A power supply is <B>required</B> for each meter/ transmitter and may be ordered separately. If using an existing power supply, order cable assembly 32704-52.
Choose transmitters with a 0 to 5 VDC or 4 to 20 mA output for control or datalogging purposes. Models 32718-00 to -09 also feature a pulse output that is proportional to flow rate.