Hedland Flow Meter, 1/4" Alum, 1.0/10 scfm for Air with Extended Cap and Gauge
Supplier: VWR International
VWR Item#MFLX32510-59
Your Organization's Price:
Retrieving
Direct read 360° rotatable scale.
  • Easy-to-read linear scale
FEATURES <br />Direct readings <br />Install in any position <br />360° Rotatable Guard/Scale <br />No Flow Straighteners or Special Piping Required <br />Insensitive to Shock and Vibration <br />Good Viscosity Stability <br />Temperature up to 240 °F <br />Accuracy +/-2% Full Scale <br />Repeatability +/-1% <br />Special Scales Available