Masterflex® Fitting, Nylon, Straight, Hose Barb Union, 5/16" ID; 10/PK
Supplier: VWR International
VWR Item#MFLX30621-88
Your Organization's Price:
Retrieving
Quick and easy assembly without tools.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Ultra smooth inner surface results in fewer extractables
  • Connection Type: ⁵/₁₆" int.Ø