Masterflex® I/P® High-Performance Precision Pump Tubing, Platinum-Cured Silicone, I/P 88; 25 ft
Supplier: VWR International
Reduce the risk of particulate entrapment with smooth inner surface.
- Free Certificate of Compliance is available - request in cart
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Single-use custom lengths and configurations available to minimize downtime
- No leachable additives protects sensitive and costly fluids
- I/P® 89 tubing has a 5/8” (15.88 mm) inner diameter
- Accepts 5/8” hose barb for tubing connection
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications<br />Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent