Masterflex® Transfer Tubing, Flexible Vinyl, 1" ID x 1-1/4" OD; 50 Ft
Supplier: VWR International
VWR Item#MFLX96480-19
Your Organization's Price:
Retrieving
Ideal for general-purpose applications.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 1" ID x 1 1/4" OD
A good general-purpose tubing material, vinyl tubing (also known as PVC tubing) offers good chemical resistance. Its transparency allows easy visual identification of flow through the tube. This vinyl tubing's good abrasion resistance means you can use it in a laboratory, on a shop floor, or on a production bench. Vinyl tubing can be used within the temperature range of -45 to 180 °F (-43 to 82 °C).