SANI-TECH STHT-C-093-3H Platinum-Cured Silicone Tubing, 3/32"×9/32"; 100 Ft/Pk
Supplier: ST. GOBAIN - TAUNTON
VWR Item#MFLX95920-55
Your Organization's Price:
Retrieving
Ultimate product traceability.
  • Resist temperature extremes, ozone, corona, radiation, moisture, compression sets, weathering, and chemical attacks
  • 1/16" ID and larger have part numbers and lot codes laser marked for permanent traceability
  • Identify tubing by external labeling too—both bag and box are marked
Applications: sterile fill and transfer, media processing, sanitary peristaltic pump tubing, general laboratory use, cell harvesting, and liquid chromatography.<br />Characteristics: withstands repeated autoclaving, ultra-flexible, smooth surface for low adhesion, tasteless, odorless, nontoxic, and translucent.<br />Certifications: Elastomer is FDA-compliant and meets USP Class VI, EP 3.1.9, and ISO 10993 biocompatibility criteria. Tubing is manufactured and packaged in a certified cleanroom.<br />Temperature range: -80 to 500 °F (-62 to 260 °C).<br />Sterilize: Autoclave, ethylene oxide, or gamma irradiation (up to 5.0 Mrad).