Double-walled PTFE heat-shrink tubing, 0.240"ID, 12 ft/pack
Supplier: VWR International
Ideal for chemically inert applications.
Double-walled PTFE tubing combines an exterior of heat-shrink PTFE with an inner layer of FEP to provide a tight, moisture-proof bond. Comes in 4-ft lengths; 3 lengths per pack, for a total of 12 ft/pk. Opaque.