Masterflex® Transfer Tubing, Platinum-Cured Silicone, 5/16" ID x 7/16" OD; 25 Ft
Supplier: VWR International
VWR Item#MFLX95802-15
Your Organization's Price:
Retrieving
No need to worry about the heat - use for applications up to 460 °F.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 5/16" ID x 7/16" OD
Applications: Pharmaceutical, biotechnology, aerospace, food and beverage, and general lab applications. <br />Characteristics: Platinum-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent. <br />Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI; exceeds 3A Sanitary Standards. Tubing is manufactured according to GMP. <br />Temperature range: -58 to 446 °F (-50 to 230 °C)<br />Sterilize: Autoclave or gamma irradiation.