Masterflex® Smart Gasket® Sanitary Sampling Gasket, Silicone, 1 Port, 1" Tri-Clamp; 1/EA
Supplier: VWR International
VWR Item#MFLX30542-08
Your Organization's Price:
Retrieving
Smart Gasket lets you take samples or readings from a sanitary line.
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.