PendoTech Single-Use 3-Way Stopcock, Polysulfone, Male/Female Luer Inlet×Female Luer Branch
Supplier: PENDOTECH
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
- Produces a 4-20 milliamp signal that is linear with temperature
- Connections to the transmitter are quick and convenient
- Convenient mounting screws
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Connects flow through tubing to luer fitting single use pressure sensors.
***Must order reusable temperature sensor cables (19406-14 for luer sensor and 19406-15 for hose barb sensors) separately.***
<b>Delivery information: </b>Power cord and plug.