Pure-Fit® TC Plastic Tubing Clamp, PVDF, 3/8"; 10/Pk
Supplier: Avantor
Install easily over existing tubing lines or assemblies.
- Smooth contours eliminate risk of puncture or rupture
- Fully sterilizable
- Meet USP Class IV criteria
The Pure-Fit TC clamp has a press-down locking mechanism that provides complete fluid stoppage. Its side-release mechanism ensures against unwanted opening. The open design can be installed over existing fittings or finished tubing assemblies, eliminating costly downtime. Available sizes handle two tubing ranges (up to 3⁄8" or 3⁄8" to 3⁄4"). Available in polypropylene (PP) or PVDF. Sterilize PP by gamma irradiation only; PVDF may be sterilized by gamma irradiation or autoclaving.